missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzenesulfonic Acid, TRC

Product Code. 30402580
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30402580 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402580 Supplier Toronto Research Chemicals Supplier No. TRCH710120100G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydrazinobenzenesulfonic Acid
CAS 98-71-5
Molecular Formula C6 H8 N2 O3 S
Quantity 100 g
Synonym p-Hydrazinobenzenesulfonic Acid, 4-Sulfophenylhydrazine, NSC 1608, Phenylhydrazine-4-sulfonic Acid, Phenylhydrazine-p-sulfonic Acid, Sulfanilic Acid Hydrazine, p-Hydrazinobenzenesulfonic Acid, p-Hydrazinophenylsulfonic Acid, p-Sulfophenylhydrazine
InChI Formula InChI=1S/C6H8N2O3S/c7-8-5-1-3-6(4-2-5)12(9,10)11/h1-4,8H,7H2,(H,9,10,11)
SMILES NNc1ccc(cc1)S(=O)(=O)O
IUPAC Name 4-hydrazinylbenzenesulfonic acid
Molecular Weight (g/mol) 188.2
Formula Weight 188.0256
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.