missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzoic Acid Hydrochloride, TRC

Product Code. 30396920
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396920 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396920 Supplier Toronto Research Chemicals Supplier No. TRCH71657510G
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydrazinobenzoic Acid Hydrochloride
CAS 24589-77-3
Molecular Formula C7 H8 N2 O2 . H Cl
Quantity 10 g
InChI Formula InChI=1S/C7H8N2O2.ClH/c8-9-6-3-1-5(2-4-6)7(10)11;/h1-4,9H,8H2,(H,10,11);1H
SMILES Cl.NNc1ccc(cc1)C(=O)O
IUPAC Name 4-hydrazinobenzoic acid;hydrochloride
Molecular Weight (g/mol) 188.612
Formula Weight 188.0353
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.