missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzoic Acid, TRC

Product Code. 30406870
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406870 5 g
30392052 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30406870 Supplier Toronto Research Chemicals Supplier No. TRCH7100205G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-67-0
Molecular Formula C7 H8 N2 O2
Molecular Weight (g/mol) 152.15
Synonym Benzoic acid, 4-hydrazinyl-, Benzoic acid, 4-hydrazino- (9CI), Benzoic acid, p-hydrazino- (6CI,7CI,8CI), (4-Carboxyphenyl)hydrazine, (p-Carboxyphenyl)hydrazine, 4-Hydrazinobenzoic acid, 4-Hydrazinylbenzoic acid, NSC 3813, p-Hydrazinobenzoic acid, Deferasirox Imp. F (EP)
IUPAC Name 4-hydrazinylbenzoic acid
SMILES NNc1ccc(cc1)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H8N2O2/c8-9-6-3-1-5(2-4-6)7(10)11/h1-4,9H,8H2,(H,10,11)
Formula Weight 152.0586
Chemical Name or Material 4-Hydrazinobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.