missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2(5H)-furanone, 95%

Product Code. 10216290 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10216290 1 g Glass bottle 1g
10209743 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10216290 Supplier Thermo Scientific Acros Supplier No. 235260010
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 541-57-1 | C4H4O3 | 100.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-57-1
Molecular Formula C4H4O3
Molecular Weight (g/mol) 100.07
MDL Number MFCD00052187
InChI Key JZQBAGOECGRTSA-UHFFFAOYSA-N
Synonym 4-hydroxyfuran-2 5h-one, 4-hydroxy-2,5-dihydrofuran-2-one, 4-hydroxy-2 5h-furanone, 2 5h-furanone, 4-hydroxy, 4-hydroxy-5h-furan-2-one, 5-hydroxyfuran-3-one, 5-hydroxyfuran-3 2h-one, pubchem21770, acmc-209xna, tetronic acid 5g
PubChem CID 54683813
IUPAC Name 3-hydroxy-2H-furan-5-one
SMILES OC1=CC(=O)OC1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 135°C to 137°C
Color Beige-Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,acetonitrile,dmso
Formula Weight 100.07
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2(5H)-furanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.