missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-butanone, 95%

Product Code. 11397776
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397776 25 g 25g
11307786 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397776 Supplier Thermo Scientific Alfa Aesar Supplier No. L11456.14
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 590-90-9 | C4H8O2 | 88.106 g/mol

4-Hydroxy-2-butanone is used in the preparation of fused benzazepine molecules as selective D3 receptor antagonists with pharmaceutical activity. It is also used in the preparation of verrucarin and mevalonic acid lactone. Further, it is used to prepare 3-buten-2-one through dehydration over an anatase titanium dioxide catalyst. It is also employed in the preparation of 4-methyl-5-hydroxymethylthiazole, (±)-lineatin and beta-hydroxylactones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-2-butanone is used in the preparation of fused benzazepine molecules as selective D3 receptor antagonists with pharmaceutical activity. It is also used in the preparation of verrucarin and mevalonic acid lactone. Further, it is used to prepare 3-buten-2-one through dehydration over an anatase titanium dioxide catalyst. It is also employed in the preparation of 4-methyl-5-hydroxymethylthiazole, (±)-lineatin and beta-hydroxylactones.

Solubility
Miscible with water, alcohol, ethanol and ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-90-9
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00059005
InChI Key LVSQXDHWDCMMRJ-UHFFFAOYSA-N
Synonym 4-hydroxy-2-butanone, 2-butanone, 4-hydroxy, methylolacetone, 3-oxo-1-butanol, 3-oxobutanol, 3-ketobutan-1-ol, monomethylolacetone, 4-butanol-2-one, unii-tcm0bj44mf, 2-hydroxyethyl methyl ketone
PubChem CID 111509
ChEBI CHEBI:41268
IUPAC Name 4-hydroxybutan-2-one
SMILES CC(=O)CCO

Specifications

Density 1.02
Boiling Point 73°C to 76°C (12 mmHg)
Flash Point 89°C (192°F)
Refractive Index 1.431
Quantity 25 g
Beilstein 1737666
Solubility Information Miscible with water,alcohol,ethanol and ether.
Formula Weight 88.11
Percent Purity 95%
Chemical Name or Material 4-Hydroxy-2-butanone
compliance-icons
Product Identifier
  • 4-Hydroxy-2-butanone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Hydroxy-2-butanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.