missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxy-3'-(trifluoromethyl)acetophenone, 95%, Thermo Scientific Chemicals

Product Code. 15444307
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15444307 1 g 1g
15497865 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444307 Supplier Thermo Scientific Alfa Aesar Supplier No. H31691.03
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 149105-11-3 | Molecular Formula: C9H7F3O2 | Molecular Weight: 204.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149105-11-3
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD01091005
InChI Key HKRUXZJKSFFSGF-UHFFFAOYSA-N
Synonym 4-hydroxy-3-trifluoromethyl acetophenone, 1-4-hydroxy-3-trifluoromethyl phenyl ethanone, 4'-hydroxy-3'-trifluoromethyl acetophenone, 1-4-hydroxy-3-trifluoromethyl phenyl ethan-1-one, 1-4-hydroxy-3-trifluoromethylphenyl ethanone, ethanone,1-4-hydroxy-3-trifluoromethyl phenyl, acmc-1bxyq, intermediates-zcf02143, 5-acetyl-2-hydroxybenzotrifluoride
PubChem CID 2775109
IUPAC Name 1-[4-hydroxy-3-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC(=C(C=C1)O)C(F)(F)F

Specifications

Melting Point 174°C to 177°C
Quantity 1 g
Formula Weight 204.15
Percent Purity 95%
Chemical Name or Material 4'-Hydroxy-3'-(trifluoromethyl)acetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.