missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-4-phenylpiperidine, 99%

Product Code. 15471648
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471648 5 g 5g
15461648 1 g 1g
15481648 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15471648 Supplier Thermo Scientific Alfa Aesar Supplier No. H55418.06
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 40807-61-2 | C11H15NO | 177.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40807-61-2
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00006000
InChI Key KQKFQBTWXOGINC-UHFFFAOYSA-N
PubChem CID 96387
IUPAC Name 4-phenylpiperidin-4-ol
SMILES C1CNCCC1(C2=CC=CC=C2)O

Specifications

Melting Point 157°C to 161°C
Color Brown
Quantity 5 g
Beilstein 141577
Synonym 4-hydroxy-4-phenylpiperidine, 4-phenyl-4-piperidinol, 4-piperidinol, 4-phenyl, 4-phenyl-piperidin-4-ol, 4phenyl-4-piperidinol, 4-phenyl-4-piperidol, acmc-1amcr, 4-piperidinol,4-phenyl, 4-phenyl-piperidine-4-ol, 4-hydroxy-4phenylpiperidine
Formula Weight 177.25
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 4-Hydroxy-4-phenylpiperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.