missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-5-iodopyrimidine, 95%, Thermo Scientific Chemicals

Product Code. 15437698
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437698 1 g 1g
15447698 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437698 Supplier Thermo Scientific Alfa Aesar Supplier No. H61190.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4349-07-9 | Molecular Formula: C4H3IN2O | Molecular Weight: 220.23 g/mol

4-Hydroxy-5-iodopyrimidine is an intermediate used in pharmaceuticals and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-5-iodopyrimidine is an intermediate used in pharmaceuticals and chemical research.

Solubility
Soluble in water (67 g/L) (25°C).

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 4349-07-9

Especificaciones

Molecular Formula C4H3IN2O
MDL Number MFCD02666079
Quantity 1 g
Sensitivity Light sensitive
Synonym 5-iodopyrimidin-4-ol, 5-iodopyrimidin-4 3h-one, 4-hydroxy-5-iodopyrimidine, 5-iodo-1h-pyrimidin-4-one, 4 1h-pyrimidinone, 5-iodo, 5-iodopyrimidin-4 1h-one, 5-iodo-3h-pyrimidin-4-one, 5-iodo-3,4-dihydropyrimidin-4-one, 5-iodopyrimidin-4-ol;5-iodopyrimidin-4 1h-one
Solubility Information Soluble in water (67g/L) (25°C).
InChI Key SMIGOGPUTQXOKF-UHFFFAOYSA-N
SMILES C1=C(C(=O)NC=N1)I
IUPAC Name 5-iodo-1H-pyrimidin-6-one
Molecular Weight (g/mol) 221.99
PubChem CID 12356582
Formula Weight 221.98
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Hydroxy-5-iodopyrimidine
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.