missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-methoxymethyl-2-(methylthio)pyrimidine, 97%, Thermo Scientific™

Product Code. 10723453
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10723453

Brand: Thermo Scientific Maybridge RJF00491EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 68087-13-8
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.229
MDL Number MFCD00052618
InChI Key OWIFCBBLFPQMSJ-UHFFFAOYSA-N
Synonym 4-hydroxy-6-methoxymethyl-2-methylthio pyrimidine, 6-methoxymethyl-2-methylthio pyrimidin-4 3h-one, 6-methoxymethyl-2-methylthio pyrimidin-4-ol, 6-methoxymethyl-2-methylsulfanyl pyrimidin-4-ol, maybridge1_006914, 6-hydroxy-4-methoxymethyl-2-methylthiopyrimidine, 6-methyloxy methyl-2-methylthio-4-pyrimidinol, 4 1h-pyrimidinone, 6-methoxymethyl-2-methylthio
PubChem CID 2730176
IUPAC Name 6-(methoxymethyl)-2-methylsulfanyl-1H-pyrimidin-4-one
SMILES COCC1=CC(=O)N=C(N1)SC

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 186.23
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-6-methoxymethyl-2-(methylthio)pyrimidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.