missing translation for 'onlineSavingsMsg'
Learn More

4’-Hydroxy Atomoxetine beta-D-Glucuronide , TRC

Product Code. 30011941
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011941 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30011941 Supplier Toronto Research Chemicals Supplier No. TRCH8025101MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 540729-08-6
Molecular Formula C23H29NO8
Quantity 1 mg
Synonym β-D-Glucopyranosiduronic acid, 3-methyl-4-[(1R)-3-(methylamino)-1-phenylpropoxy]phenyl, 3-Methyl-4-[(1R)-3-(methylamino)-1-phenylpropoxy]phenyl β-D-glucopyranosiduronic acid, 4-Hydroxyatomoxetine-O-glucuronide
InChI Formula InChI=1S/C23H29NO8/c1-13-12-15(30-23-20(27)18(25)19(26)21(32-23)22(28)29)8-9-16(13)31-17(10-11-24-2)14-6-4-3-5-7-14/h3-9,12,17-21,23-27H,10-11H2,1-2H3,(H,28,29)/t17-,18+,19+,20-,21+,23-/m1/s1
SMILES CNCC[C@@H](Oc1ccc(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)cc1C)c3ccccc3
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[3-methyl-4-[(1R)-3-(methylamino)-1-phenyl-propoxy]phenoxy]tetrahydropyran-2-carboxylic acid
Molecular Weight (g/mol) 447.48
Formula Weight 447.1893
Chemical Name or Material 4'-Hydroxy Atomoxetine beta-D-Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.