missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-TEMPO, free radical, 98+%

Product Code. 11434134
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11434134

Brand: Thermo Scientific Alfa Aesar A12497.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2226-96-2 | C9H18NO2

Superoxide scavenger with neuroprotective, anti-inflammatory and analgesic effects4-Hydroxy-TEMPO free radical is used as a catalyst and a chemical oxidant by virtue of being a stable radical. It is used for detoxifying reactive oxygen species in medicinal chemistry. It is involved in the catalysis of disproportionation of superoxide and facilitates hydrogen peroxide metabolism. It is used to prepare tris(2,2,6,6-tetramethyl-1-oxyl-4-piperidyloxy)trimellitate by reacting with benzene-1,2,4-tricarbonyl trichloride. Further, it is used as a free radical scavenger and exhibits neuroprotective, anti-inflammatory and analgesic effects. In addition to this, it has good inhibition effect to acrylic acid esters, methyl acrylate, acrylic acid, acrylonitrile, styrene and butadiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Superoxide scavenger with neuroprotective, anti-inflammatory and analgesic effects4-Hydroxy-TEMPO free radical is used as a catalyst and a chemical oxidant by virtue of being a stable radical. It is used for detoxifying reactive oxygen species in medicinal chemistry. It is involved in the catalysis of disproportionation of superoxide and facilitates hydrogen peroxide metabolism. It is used to prepare tris(2,2,6,6-tetramethyl-1-oxyl-4-piperidyloxy)trimellitate by reacting with benzene-1,2,4-tricarbonyl trichloride. Further, it is used as a free radical scavenger and exhibits neuroprotective, anti-inflammatory and analgesic effects. In addition to this, it has good inhibition effect to acrylic acid esters, methyl acrylate, acrylic acid, acrylonitrile, styrene and butadiene.

Solubility
Soluble in water, ethanol, methanol and dimethyl sulfoxide.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2226-96-2
Molecular Formula C9H18NO2
MDL Number MFCD00006478
Synonym 4-Hydroxy-2,2,6,6-tetramethyl-1-piperidin-1-yloxy, free radical

Specifications

Melting Point 69°C to 72°C
pH 8.2
Flash Point 146°C (295°F)
Quantity 5 g
Beilstein 1422990
Solubility Information Soluble in water,ethanol,methanol and dimethyl sulfoxide.
Formula Weight 172.25
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-TEMPO, free radical
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Hydroxy-TEMPO, free radical
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-hydroxy-2,2,6,6-tetramethyl-1-piperidinyloxy

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.