missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Valsartan (Mixture of Diastereomers), TRC

Product Code. 30398935
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398935 25 mg -
30402925 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30398935 Supplier Toronto Research Chemicals Supplier No. TRCH99500025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188259-69-0
Molecular Formula C24 H29 N5 O4
Molecular Weight (g/mol) 451.52
Synonym 4-Hydroxy Valsartan
IUPAC Name (2S)-2-[4-hydroxypentanoyl-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]amino]-3-methylbutanoic acid
SMILES CC(O)CCC(=O)N(Cc1ccc(cc1)c2ccccc2c3nnn[nH]3)[C@@H](C(C)C)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C24H29N5O4/c1-15(2)22(24(32)33)29(21(31)13-8-16(3)30)14-17-9-11-18(12-10-17)19-6-4-5-7-20(19)23-25-27-28-26-23/h4-7,9-12,15-16,22,30H,8,13-14H2,1-3H3,(H,32,33)(H,25,26,27,28)/t16?,22-/m0/s1
Formula Weight 451.222
Chemical Name or Material 4-Hydroxy Valsartan, Mixture of Diastereomers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.