missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxyacetophenone, 99%

Product Code. 11314719
Change view
Click to view available options
Quantity:
100 g
500 g
1000 g
Unit Size:
1000g
100g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314719 100 g 100g
11324719 500 g 500g
11404004 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11314719 Supplier Thermo Scientific Alfa Aesar Supplier No. A11974.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-93-4 | C8H8O2 | 136.15 g/mol

4?-Hydroxyacetophenone has been used as ketone component in the preparation of 1-aryl-3-phenethylamino-1-propanone hydrochlorides, potential cytotoxic agents, via Mannich reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Hydroxyacetophenone has been used as ketone component in the preparation of 1-aryl-3-phenethylamino-1-propanone hydrochlorides, potential cytotoxic agents, via Mannich reactions.

Solubility
Soluble in 95% Ethanol. Slightly soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-93-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002359
InChI Key TXFPEBPIARQUIG-UHFFFAOYSA-N
Synonym 4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone
PubChem CID 7469
ChEBI CHEBI:28032
IUPAC Name 1-(4-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 106°C to 108°C
Boiling Point 286°C
Flash Point 166°C (330°F)
Quantity 100 g
Beilstein 774355
Solubility Information Soluble in 95% Ethanol. Slightly soluble in water.
Formula Weight 136.15
Percent Purity 99%
Chemical Name or Material 4'-Hydroxyacetophenone
compliance-icons
Identifiant produit
  • 4'-Hydroxyacetophenone
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Informations supplémentaires
  • MIXTURE LIST-Contain: p-Hydroxyacetophenone

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.