missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybutanesulfonate Sodium Salt (Technical Grade), TRC

Product Code. 30007989
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007989 5 g 5g
30011470 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007989 Supplier Toronto Research Chemicals Supplier No. TRCH9535955G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31465-25-5
Molecular Formula C4 H9 O4 S . Na
Molecular Weight (g/mol) 176.17
Synonym 4-Hydroxy-1-butanesulfonic Acid Sodium Salt (1:1), 1-Butanesulfonic acid, 4-hydroxy-, monosodium salt (8CI,9CI), Sodium 4-hydroxybutanesulfonate, WAS 17
IUPAC Name sodium;4-hydroxybutane-1-sulfonate
SMILES [Na+].OCCCCS(=O)(=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4H10O4S.Na/c5-3-1-2-4-9(6,7)8;/h5H,1-4H2,(H,6,7,8);/q;+1/p-1
Formula Weight 176.0119
Chemical Name or Material 4-Hydroxybutanesulfonate sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.