missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybutyl acrylate, 95%, stab. with ca 500ppm 4-methoxyphenol

Product Code. 11387546
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387546 25 g 25g
11377546 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387546 Supplier Thermo Scientific Alfa Aesar Supplier No. L11097.14
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2478-10-6 | C7H16ClNO2S | 213.72 g/mol

4-Hydroxybutyl acrylate is used in paint and coating materials (excellent scratch resistance, mechanical properties, and acid rain resistance ), pressure sensitive adhesives, UV/EB curable composition, waterborne coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybutyl acrylate is used in paint and coating materials (excellent scratch resistance, mechanical properties, and acid rain resistance ), pressure sensitive adhesives, UV/EB curable composition, waterborne coatings.

Solubility
Fully miscible in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2478-10-6
MDL Number MFCD00010261

Specifications

Melting Point -112°C
Density 1.041
Boiling Point ∼230°C
Flash Point >110°C (230°F)
Molecular Formula C7H16ClNO2S
Refractive Index 1.452
Quantity 25 g
UN Number UN3082
Beilstein 1756545
Synonym s-tert-butyl-l-cysteine hydrochloride, h-cys tbu-oh.hcl, h-cys tbu-oh hcl, s-t-butyl-l-cysteine hydrochloride, h-cys tbu-ohhcl, s-tert-butyl-l-cysteine-hydrogen chloride 1/1, 2r-2-amino-3-tert-butylsulfanyl propanoic acid hydrochloride, pubchem19026, h-cys but-oh hcl, h-cys but-oh hydrochloride
Solubility Information Fully miscible in water.
InChI Key MHBMYFJKEBCMDR-JEDNCBNOSA-N
SMILES CC(C)(C)SCC(C(=O)O)N.Cl
IUPAC Name (2R)-2-amino-3-tert-butylsulfanylpropanoic acid;hydrochloride
Molecular Weight (g/mol) 213.72
PubChem CID 17750295
Formula Weight 144.17
Percent Purity 95%
Chemical Name or Material 4-Hydroxybutyl acrylate, stabilized with ≈500ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.