missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyindole, TRC

Product Code. 30389941
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389941 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389941 Supplier Toronto Research Chemicals Supplier No. TRCH943515500MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1H-Indol-4-ol
CAS 2380-94-1
Molecular Formula C8 H7 N O
Quantity 500 mg
Synonym 1H-Indol-4-ol, Pindolol Imp. E (EP), 4-Hydroxyindole
InChI Formula InChI=1S/C8H7NO/c10-8-3-1-2-7-6(8)4-5-9-7/h1-5,9-10H
SMILES Oc1cccc2[nH]ccc12
IUPAC Name 1H-indol-4-ol
Molecular Weight (g/mol) 133.15
Formula Weight 133.0528
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.