missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxymethyl-5-methylimidazole, TRC

Product Code. 30389761
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389761 500 mg -
30407898 50 mg 50mg
30405497 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389761 Supplier Toronto Research Chemicals Supplier No. TRCH005070500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29636-87-1
Molecular Formula C5 H8 N2 O
Molecular Weight (g/mol) 112.13
Synonym Cimetidine EP Impurity I
IUPAC Name (4-methyl-1H-imidazol-5-yl)methanol
SMILES Cc1nc[nH]c1CO

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C5H8N2O/c1-4-5(2-8)7-3-6-4/h3,8H,2H2,1H3,(H,6,7)
Formula Weight 112.064
Chemical Name or Material 4-Hydroxymethyl-5-methylimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.