missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)imidazole hydrochloride, 99%, Thermo Scientific™

Product Code. 10327861 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10327861

Brand: Thermo Scientific 204870010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 32673-41-9
Molecular Formula C4H7ClN2O
Molecular Weight (g/mol) 134.56
MDL Number MFCD00012697
InChI Key WFNASTYGEKUMIY-UHFFFAOYSA-N
Synonym 4-imidazolemethanol hydrochloride, 4-hydroxymethyl imidazole hydrochloride, 1h-imidazol-4-yl methanol hydrochloride, 4-hydroxymethylimidazole hydrochloride, imidazol-4-ylmethanol hydrochloride, 1h-imidazol-4-ylmethanol hydrochloride, 1h-imidazol-5-ylmethanol hydrochloride, 4-hydroxymethyl imidazole hcl, 1h-imidazole-4-ylmethanol hydrochloride, 4 5-hydroxymethylimidazole hydrochloride
PubChem CID 122926
IUPAC Name 1H-imidazol-5-ylmethanol;hydrochloride
SMILES Cl.OCC1=CN=CN1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 110°C to 112°C
Color Beige to Yellow or Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Beilstein 23, I, 103
Formula Weight 134.57
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-(Hydroxymethyl)imidazole hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Hydroxymethyl)imidazole hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.