missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylacetic Acid-d6, TRC

Product Code. 30400396
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30400396 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30400396 Supplier Toronto Research Chemicals Supplier No. TRCH949062100MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxyphenylacetic Acid-d6
CAS 100287-06-7
Molecular Formula C8 D6 H2 O3
Quantity 100 mg
Synonym 4-Hydroxybenzeneacetic-d6 Acid, (4-Hydroxyphenyl)acetic-d6 Acid, (p-Hydroxyphenyl)acetic-d6 Acid, 2-[4-(Hydroxy)phenyl]acetic-d6 Acid, 4-(Carboxymethyl)phenol, 4-Hydroxybenzeneacetic-d6 Acid, p-Hydroxybenzeneacetic-d6 Acid, (4-Hydroxyphenyl-2,3,5,6-d4)acetic-2,2-d2 Acid
InChI Formula InChI=1S/C8H8O3/c9-7-3-1-6(2-4-7)5-8(10)11/h1-4,9H,5H2,(H,10,11)/i1D,2D,3D,4D,5D2
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1O)C([2H])([2H])C(=O)O
IUPAC Name 2,2-dideuterio-2-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)acetic acid
Molecular Weight (g/mol) 158.18
Formula Weight 158.085
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.