missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-1-phenyl-1H-pyrazole, 95%

Product Code. 15405367
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15405367 250 mg 250mg
15495357 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15405367 Supplier Thermo Scientific Alfa Aesar Supplier No. H32612.MD
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 23889-85-2 | C9H7IN2 | 270.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23889-85-2
Molecular Formula C9H7IN2
Molecular Weight (g/mol) 270.073
MDL Number MFCD11707168
InChI Key PYJVDCYRDQUUKO-UHFFFAOYSA-N
Synonym 4-iodo-1-phenyl-1h-pyrazole, 1-phenyl-4-iodopyrazole, 1h-pyrazole, 4-iodo-1-phenyl, pubchem13298, 4-iodanyl-1-phenyl-pyrazole, 1-phenyl-4-iodo-1h-pyrazole
PubChem CID 14972076
IUPAC Name 4-iodo-1-phenylpyrazole
SMILES C1=CC=C(C=C1)N2C=C(C=N2)I

Specifications

Quantity 250 mg
Formula Weight 270.07
Percent Purity 95%
Chemical Name or Material 4-Iodo-1-phenyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.