missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2,1,3-benzothiadiazole, 97%, Thermo Scientific™

Product Code. 10100592
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10100592 250 mg 250mg
10394132 1 g 1g
10396914 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10100592 Supplier Thermo Scientific Maybridge Supplier No. CC09111CB
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 352018-95-2
Molecular Formula C6H3IN2S
Molecular Weight (g/mol) 262.07
MDL Number MFCD02681904
InChI Key ROPRWVSWECJQIC-UHFFFAOYSA-N
Synonym 4-iodobenzo c 1,2,5 thiadiazole, 2,1,3-benzothiadiazole,4-iodo, 4-iodo-benzo 1,2,5 thiadiazole, 4-iodanyl-2,1,3-benzothiadiazole, 4-iodobenzo c 1,2,5-thiadiazole, 4-iodo-2,1,3-benzothiadiazole
PubChem CID 2776287
IUPAC Name 4-iodo-2,1,3-benzothiadiazole
SMILES IC1=CC=CC2=NSN=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 262.07
Percent Purity 97%
Chemical Name or Material 4-Iodo-2, 1, 3-benzothiadiazole
compliance-icons
Product Identifier
  • 4-Iodo-2,1,3-benzothiadiazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Iodo-2,1,3-benzothiadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.