missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2-methyl-1H-imidazole, 97%, Thermo Scientific™

Product Code. 15219786 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15219786 25 g 25g
15209786 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15219786 Supplier Thermo Scientific Acros Supplier No. 451580250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 73746-45-9
Molecular Formula C4H5IN2
Molecular Weight (g/mol) 208.00
MDL Number MFCD02684335
InChI Key SEDSLMWYUQACGR-UHFFFAOYSA-N
PubChem CID 2794623
IUPAC Name 5-iodo-2-methyl-1H-imidazole
SMILES CC1=NC=C(I)N1

Specifications

Melting Point 138.0°C to 144.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 25 g
Synonym 4-iodo-2-methyl-1h-imidazole, 2-methyl-4 5-iodo-1 h-imidazole, 4-iodo-2-methylimidazole, 2-methyl-4-iodo-1h-imidazole, 4-iodo-2-methyl-3h-imidazole, 1h-imidazole, 4-iodo-2-methyl, 2-methyl-4-jodimidazol, ksc638k7d, 1h-imidazole,4-iodo-2-methyl, 1h-imidazole,5-iodo-2-methyl
Formula Weight 208
Percent Purity 97%
Chemical Name or Material 4-Iodo-2-methyl-1H-imidazole
compliance-icons compliance-icons
Product Identifier
  • 4-Iodo-2-methyl-1H-imidazole, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.