missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-3,5-dimethylisoxazole, 97%, Thermo Scientific™

Product Code. 11438747
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438747 5 g 5g
11428747 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438747 Supplier Thermo Scientific Alfa Aesar Supplier No. A19911.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10557-85-4
Molecular Formula C5H6INO
Molecular Weight (g/mol) 223.01
MDL Number MFCD00173743
InChI Key NMNOXVWRJISEFE-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-iodoisoxazole, 4-iodo-3,5-dimethylisoxazole, isoxazole, 4-iodo-3,5-dimethyl, 4-iodo-3,5-dimethyl-isoxazole, 3,5-dimethyl-4-iodoisorazole, pubchem3546, buttpark 97\06-60, acmc-2098gw, ksc496a2p, 3.5-dimethyl-4-iodoisorazole
PubChem CID 613883
IUPAC Name 4-iodo-3,5-dimethyl-1,2-oxazole
SMILES CC1=C(I)C(C)=NO1

Spécification

Melting Point 51°C to 55°C
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 223.01
Percent Purity 97%
Chemical Name or Material 4-Iodo-3,5-dimethylisoxazole
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.