missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-5-methyl-3-phenylisoxazole, 97%, Thermo Scientific™

Product Code. 10049201
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10049201 1 g 1g
10582842 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10049201 Supplier Thermo Scientific Maybridge Supplier No. CC00511DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 31295-66-6
Molecular Formula C10H8INO
Molecular Weight (g/mol) 285.084
MDL Number MFCD02681888
InChI Key DDVPBKUHWSZYIP-UHFFFAOYSA-N
Synonym 4-iodo-5-methyl-3-phenylisoxazole, isoxazole, 4-iodo-5-methyl-3-phenyl, 3-phenyl-4-iodo-5-methylisoxazole
PubChem CID 2776150
IUPAC Name 4-iodo-5-methyl-3-phenyl-1,2-oxazole
SMILES CC1=C(C(=NO1)C2=CC=CC=C2)I

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 285.08
Percent Purity 97%
Chemical Name or Material 4-Iodo-5-methyl-3-phenylisoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.