missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-L-phenylalanine, 95%

Product Code. 15458457
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15458457 5 g 5g
15468457 250 mg 250mg
15448457 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15458457 Supplier Thermo Scientific Alfa Aesar Supplier No. H51979.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24250-85-9 | C9H10INO2 | 291.09 g/mol

4-Iodo-L-phenylalanine may be used in protein engineering as a model unnatural α amino acid to alter primary amino acid composition via the opal (UGA) codon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodo-L-phenylalanine may be used in protein engineering as a model unnatural α amino acid to alter primary amino acid composition via the opal (UGA) codon.

Solubility
Partly miscible with water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24250-85-9
Molecular Formula C9H10INO2
Molecular Weight (g/mol) 291.09
MDL Number MFCD00002602
InChI Key PZNQZSRPDOEBMS-QMMMGPOBSA-N
Synonym 4-iodo-l-phenylalanine, 4-iodophenylalanine, l-4-iodophenylalanine, iodo-phenylalanine, p-iodo-l-phenylalanine, p-iodophenylalanine, s-2-amino-3-4-iodophenyl propanoic acid, 2s-2-amino-3-4-iodophenyl propanoic acid, h-phe 4-i-oh, phenylalanine, 4-iodo
PubChem CID 134497
IUPAC Name (2S)-2-amino-3-(4-iodophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(I)C=C1)C(O)=O

Specifications

Beilstein 4411317
Solubility Information Partly miscible with water.
Quantity 5 g
Formula Weight 291.09
Optical Rotation −6.5° (c=1 in Acetic Acid:Water (4:1))
Sensitivity Light sensitive
Percent Purity 95%
Chemical Name or Material 4-Iodo-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.