missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzenesulfonyl chloride, 97%

Product Code. 11495867
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495867 100 g 100g
11475867 5 g 5g
11485867 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495867 Supplier Thermo Scientific Alfa Aesar Supplier No. B24416.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 98-61-3 | C6H4ClIO2S | 302.51 g/mol

It is used as intermediate in organic syntheses & in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as intermediate in organic syntheses & in medicine.

Solubility
Reacts with water.

Notes
Moisture & light sensitive. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-61-3
Molecular Formula C6H4ClIO2S
Molecular Weight (g/mol) 302.51
MDL Number MFCD00007441
InChI Key POXFXTSTVWDWIR-UHFFFAOYSA-N
Synonym pipsyl chloride, 4-iodobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-iodo, p-iodobenzenesulfonyl chloride, 4-iodobenzenesulphonyl chloride, 4-iodobenzenesulfonylchloride, 4-iodophenylsulfonyl chloride, iodobenzene-p-sulfonyl chloride, unii-mvl274ezog, mvl274ezog
PubChem CID 7399
IUPAC Name 4-iodobenzenesulfonyl chloride
SMILES C1=CC(=CC=C1S(=O)(=O)Cl)I

Specifications

Melting Point 79°C to 84°C
Quantity 100 g
UN Number UN3261
Beilstein 2691661
Sensitivity Light and moisture sensitive
Merck Index 14,7486
Solubility Information Reacts with water.
Formula Weight 302.52
Percent Purity 97%
Chemical Name or Material 4-Iodobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.