missing translation for 'onlineSavingsMsg'
Learn More

4-(Iodomethyl)tetrahydro-2h-pyran, 97%, Thermo Scientific™

Product Code. 10686433
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10686433 1 g Amber Glass Bottle 1g
10570554 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10686433 Supplier Thermo Scientific Maybridge Supplier No. CC29940DA
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 101691-94-5
Molecular Formula C6H11IO
Molecular Weight (g/mol) 226.057
InChI Key MQLFSPBSNWUXSO-UHFFFAOYSA-N
Synonym 4-iodomethyl tetrahydro-2h-pyran, 4-iodomethyl oxane, 4-iodomethyl-tetrahydro-pyran, 4-iodomethyl tetrahydropyran, 2h-pyran, tetrahydro-4-iodomethyl, 4-iodomethyl-tetrahydro-2h-pyran, 2h-pyran,tetrahydro-4-iodomethyl, acmc-20a1ce, 4-iodomethyltetrahydropyran, 4-iodomethyl-tetrahydropyran
PubChem CID 2795507
IUPAC Name 4-(iodomethyl)oxane
SMILES C1COCCC1CI

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 226.05
Percent Purity 97%
Chemical Name or Material 4-(Iodomethyl)tetrahydro-2H-pyran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.