missing translation for 'onlineSavingsMsg'
Learn More

4-Iodopyridine, 96%

Product Code. 11407146
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11407146 1 g 1g
11417146 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11407146

Brand: Thermo Scientific Alfa Aesar B25459.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15854-87-2 | C5H4IN | 205.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15854-87-2
Molecular Formula C5H4IN
Molecular Weight (g/mol) 205.00
MDL Number MFCD02093937
InChI Key RTLUPHDWSUGAOS-UHFFFAOYSA-N
Synonym pyridine, 4-iodo, 4-iodo pyridine, 4-iodo-pyridine, p-iodopyridine, 4-iodanylpyridine, acmc-1btjc, 4-iodopyridine, ksc178q2d
PubChem CID 609492
IUPAC Name 4-iodopyridine
SMILES IC1=CC=NC=C1

Specifications

Melting Point 96°C to 99°C
Quantity 1 g
Beilstein 1594
Sensitivity Light sensitive
Formula Weight 205
Percent Purity 96%
Chemical Name or Material 4-Iodopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.