missing translation for 'onlineSavingsMsg'
Learn More

4-Iodotetrahydrothiopyran, 97%, Thermo Scientific™

Product Code. 10368823
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10368823 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10368823 Supplier Thermo Scientific Maybridge Supplier No. CC61911DA
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 281204-90-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H9IS
MDL Number MFCD09025908
Quantity 1 g
Synonym 4-iodotetrahydrothiopyran, 4-iodotetrahydro-2h-thiopyran, 2h-thiopyran,tetrahydro-4-iodo
InChI Key JVBIONNYIWEILC-UHFFFAOYSA-N
SMILES IC1CCSCC1
IUPAC Name 4-iodothiane
Molecular Weight (g/mol) 228.09
PubChem CID 18525911
Formula Weight 228.09
Percent Purity 97%
Chemical Name or Material 4-Iodotetrahydrothiopyran
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.