missing translation for 'onlineSavingsMsg'
Learn More

4-Isocyanato-1-methyl-1H-indole, 97%, Thermo Scientific™

Product Code. 10235334
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
10235334 250 mg 250mg
10502865 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 10235334

Brand: Thermo Scientific Maybridge CC45806CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 887922-92-1
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
MDL Number MFCD08690262
InChI Key TVGVHXVLMROUII-UHFFFAOYSA-N
Synonym 4-isocyanato-1-methyl-1h-indole, 1h-indole, 4-isocyanato-1-methyl, 1-methylindol-4-isocyanate, 1h-indole,4-isocyanato-1-methyl, 1-methyl-1h-indole-4-yl isocyanate
PubChem CID 18525783
IUPAC Name 4-isocyanato-1-methylindole
SMILES CN1C=CC2=C(C=CC=C21)N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 172.19
Percent Purity 97%
Chemical Name or Material 4-Isocyanato-1-methyl-1H-indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.