missing translation for 'onlineSavingsMsg'
Learn More

4-Isocyanato-1-methyl-1H-pyrazole, 95%

Product Code. p-4228750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10391710 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10391710 Supplier Thermo Scientific Acros Supplier No. 431140050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1174064-57-3 | C5H5N3O | 123.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1174064-57-3
Molecular Formula C5H5N3O
Molecular Weight (g/mol) 123.12
MDL Number MFCD14706124
InChI Key TXMLNGHIFGKEFH-UHFFFAOYSA-N
Synonym 4-isocyanato-1-methyl-1h-pyrazole
PubChem CID 45790024
IUPAC Name 4-isocyanato-1-methylpyrazole
SMILES CN1C=C(C=N1)N=C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Boiling Point 203.0°C to 205.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 123.12
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Isocyanato-1-methyl-1H-pyrazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Isocyanato-1-methyl-1H-pyrazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.