missing translation for 'onlineSavingsMsg'
Learn More

4-Isocyanato-1-methyl-1H-pyrazole, 95%

Product Code. 10391710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Pakningsstørrelse:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
10391710 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 10391710 Leverandør Thermo Scientific Acros Leverandørnr. 431140050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

CAS: 1174064-57-3 | C5H5N3O | 123.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1174064-57-3
Molecular Formula C5H5N3O
Molecular Weight (g/mol) 123.12
MDL Number MFCD14706124
InChI Key TXMLNGHIFGKEFH-UHFFFAOYSA-N
Synonym 4-isocyanato-1-methyl-1h-pyrazole
PubChem CID 45790024
IUPAC Name 4-isocyanato-1-methylpyrazole
SMILES CN1C=C(C=N1)N=C=O

Especificaciones

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Boiling Point 203.0°C to 205.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 123.12
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Isocyanato-1-methyl-1H-pyrazole
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • 4-Isocyanato-1-methyl-1H-pyrazole
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.