missing translation for 'onlineSavingsMsg'
Learn More

4-Isocyanatobenzeneboronic acid pinacol ester, 95%

Product Code. 15480286
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15480286 250 mg 250mg
15439777 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15480286 Supplier Thermo Scientific Alfa Aesar Supplier No. H53003.MD
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 380430-64-8 | C13H16BNO3 | 245.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380430-64-8
Molecular Formula C13H16BNO3
Molecular Weight (g/mol) 245.085
MDL Number MFCD03411942
InChI Key RHKRIDVZGAXYQE-UHFFFAOYSA-N
Synonym 2-4-isocyanatophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-isocyanatophenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-4-isocyanatophenyl-4,4,5,5-tetramethyl, 4-isocyanatophenylboronic acid, pinacol ester, 4-isocyanatophenyl boronic acid pinacol ester, 4-isocyanatobenzeneboronic acid pinacol ester, 4-isocyanatophenyl boronic acid, pinacol ester, 4-isocyanatophenyl boronic acid,pinacol ester, 2-4-isocyanato-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane
PubChem CID 2773474
IUPAC Name 2-(4-isocyanatophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N=C=O

Specifications

Melting Point 162°C to 168°C
Quantity 250 mg
Formula Weight 245.08
Percent Purity 95%
Chemical Name or Material 4-Isocyanatobenzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.