missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropoxyphenylboronic acid, 97%, Thermo Scientific™

Product Code. 10244734 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10244734 1 g Glass bottle 1g
10461484 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10244734 Supplier Thermo Scientific Acros Supplier No. 393930010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 153624-46-5
Molecular Formula C9H13BO3
Molecular Weight (g/mol) 180.01
MDL Number MFCD03427051
InChI Key CJUHQADBFQRIMC-UHFFFAOYSA-N
Synonym 4-isopropoxyphenylboronic acid, 4-isopropoxylphenylboronic acid, 4-isopropoxyphenyl boronic acid, 4-isopropoxybenzeneboronic acid, 4-isopropyloxyphenylboronic acid, 4-propan-2-yloxyphenyl boronic acid, boronic acid, 4-1-methylethoxy phenyl, 4-propan-2-yloxy phenyl boronic acid, 4-iso-propoxyphenylboronic acid, p-isopropoxyphenylboronic acid
PubChem CID 3698726
IUPAC Name (4-propan-2-yloxyphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)OC(C)C)(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Tan or White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3)2CHOC6H4B(OH)2
Quantity 1 g
Formula Weight 180.01
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Isopropoxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Isopropoxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.