missing translation for 'onlineSavingsMsg'
Learn More

4'-Isopropylacetophenone, 96%, ACROS Organics™

Product Code. 10342193 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10342193 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10342193 Supplier Thermo Scientific Acros Supplier No. 381830050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 645-13-6
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C11H14O
Linear Formula (CH3)2CHC6H4COCH3
Refractive Index 1.5220 to 1.5240 (20°C, 589nm)
MDL Number MFCD00048297
Quantity 5 g
Synonym 4'-isopropylacetophenone, 4-isopropylacetophenone, p-isopropylacetophenone, cuminone, 1-4-isopropylphenyl ethanone, ethanone, 1-4-1-methylethyl phenyl, acetophenone, 4'-isopropyl, unii-742zkz2a6d, methyl p-isopropylphenyl ketone, 1-4-propan-2-yl phenyl ethan-1-one
InChI Key PDLCCNYKIIUWHA-UHFFFAOYSA-N
SMILES CC(C)C1=CC=C(C=C1)C(C)=O
IUPAC Name 1-[4-(propan-2-yl)phenyl]ethan-1-one
Molecular Weight (g/mol) 162.23
PubChem CID 12578
Formula Weight 162.23
Percent Purity ≥95%
Physical Form Liquid
Chemical Name or Material 4'-Isopropylacetophenone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.