missing translation for 'onlineSavingsMsg'
Learn More

4'-Isopropylacetophenone, 97%

Artikelnummer. 11468977
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
5 g
25 g
100 g
Pakningsstørrelse:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
11468977 100 g 100g
11448977 5 g 5g
11458977 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11468977 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. B20581.22
 Restricted Item
Estimated Shipment: 02-12-2026
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 645-13-6 | C11H14O | 162.23 g/mol

4'-Isopropylacetophenone is used as perfuming agents. It is also used as flavoring and fragrance agent. Indenes are yielded from the derivatives if this compound in one step synthesis using a heterogeneous catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Isopropylacetophenone is used as perfuming agents. It is also used as flavoring and fragrance agent. Indenes are yielded from the derivatives if this compound in one step synthesis using a heterogeneous catalyst.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 645-13-6
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00048297
InChI Key PDLCCNYKIIUWHA-UHFFFAOYSA-N
Synonym 4'-isopropylacetophenone, 4-isopropylacetophenone, p-isopropylacetophenone, cuminone, 1-4-isopropylphenyl ethanone, ethanone, 1-4-1-methylethyl phenyl, acetophenone, 4'-isopropyl, unii-742zkz2a6d, methyl p-isopropylphenyl ketone, 1-4-propan-2-yl phenyl ethan-1-one
PubChem CID 12578
IUPAC Name 1-(4-propan-2-ylphenyl)ethanone
SMILES CC(C)C1=CC=C(C=C1)C(C)=O

Tekniske data

Density 0.97
Boiling Point 238°C
Refractive Index 1.523
Quantity 100 g
Beilstein 2205694
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 162.23
Percent Purity 98%
Chemical Name or Material 4'-Isopropylacetophenone

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.