missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzoic acid, 98+%

Product Code. 10607854 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10607854 25 g Glass bottle 25g
10538902 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10607854 Supplier Thermo Scientific Acros Supplier No. 412800250
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 536-66-3 | C10H12O2 | 164.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-66-3
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00002564
InChI Key CKMXAIVXVKGGFM-UHFFFAOYSA-N
Synonym 4-isopropylbenzoic acid, cuminic acid, cumic acid, p-isopropylbenzoic acid, 4-1-methylethyl benzoic acid, benzoic acid, 4-1-methylethyl, 4-propan-2-yl benzoic acid, benzoic acid, p-isopropyl, p-cumic acid, 4-isopropyl benzoic acid
PubChem CID 10820
ChEBI CHEBI:28122
IUPAC Name 4-propan-2-ylbenzoic acid
SMILES CC(C)C1=CC=C(C=C1)C(=O)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 116.0°C to 120.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3)2CHC6H4COOH
Quantity 25 g
Merck Index 15, 2608
Solubility Information Solubility in water: sparingly soluble
Formula Weight 164.2
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 4-Isopropylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Isopropylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.