missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzyl chloride, 95%

Product Code. 11312657
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11312657

Brand: Thermo Scientific Alfa Aesar L11839.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2051-18-5 | C10H13Cl | 168.664 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-18-5
Molecular Formula C10H13Cl
Molecular Weight (g/mol) 168.664
MDL Number MFCD00018885
InChI Key CYAKWEQUWJAHLW-UHFFFAOYSA-N
Synonym 4-isopropylbenzyl chloride, 1-chloromethyl-4-isopropylbenzene, p-cymene, 7-chloro, p-isopropylbenzyl chloride, 7-chloro-p-cymene, 4-isopropylbenzylchloride, benzene, 1-chloromethyl-4-1-methylethyl, 1-chloromethyl-4-propan-2-yl benzene, p-isopropyl benzyl chloride, benzene, 1-chloromethyl-4-methylethyl
PubChem CID 74916
IUPAC Name 1-(chloromethyl)-4-propan-2-ylbenzene
SMILES CC(C)C1=CC=C(C=C1)CCl

Specifications

Density 1.02
Boiling Point 119°C to 120°C (35 mmHg)
Flash Point 104°C (219°F)
Odor Pungent
Refractive Index 1.523
Quantity 5 g
UN Number UN3265
Beilstein 907207
Formula Weight 168.67
Percent Purity 95%
Chemical Name or Material 4-Isopropylbenzyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Isopropylbenzyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-Isopropylbenzyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.