missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylcyclohexanone, 96%

Product Code. 11392756
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11392756

Brand: Thermo Scientific Alfa Aesar L08863.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5432-85-9 | C9H16O | 140.23 g/mol

4-Isopropylcyclohexanone is used to prepare 4-isopropyl-cyclohexanone oxime. It is also used in the synthesis of dihydroindol-2-ones as agonists and antagonist ligands at the nociceptin receptor. Further, it is used in the preparation of beta-alanine derivatives, which acts as orally available glucagon receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Isopropylcyclohexanone is used to prepare 4-isopropyl-cyclohexanone oxime. It is also used in the synthesis of dihydroindol-2-ones as agonists and antagonist ligands at the nociceptin receptor. Further, it is used in the preparation of beta-alanine derivatives, which acts as orally available glucagon receptor antagonists.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Incompatible with oxidizing agents. reducing agents and plastics.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5432-85-9
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00043479
InChI Key FPKISACHVIIMRA-UHFFFAOYSA-N
Synonym 4-isopropylcyclohexanone, cyclohexanone, 4-1-methylethyl, 4-propan-2-yl cyclohexan-1-one, 4-isopropylcyclohexan-1-one, cyclohexanone, 4-isopropyl, 4-isopropyl-cyclohexanone, 4-iso-propylcyclohexanone, 4-methylethyl cyclohexan-1-one, acmc-20a5fk, 4-isopropyl cyclohexanone
PubChem CID 79488
IUPAC Name 4-propan-2-ylcyclohexan-1-one
SMILES CC(C)C1CCC(=O)CC1

Specifications

Boiling Point 94°C to 96°C (17 mmHg)
Flash Point 80°C (176°F)
Refractive Index 1.458
Quantity 2 g
Beilstein 1236952
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 140.23
Percent Purity 96%
Chemical Name or Material 4-Isopropylcyclohexanone

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado