missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylthioxanthone, TRC

Product Code. 17984335
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17984335 50 mg 50mg
17961345 100 mg 100mg
17941785 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17984335 Supplier Toronto Research Chemicals Supplier No. TRCI87350550MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83846-86-0
Molecular Formula C16 H14 O S
Molecular Weight (g/mol) 254.35
Synonym 4-(1-Methylethyl)-9H-thioxanthen-9-one, 4-Isopropyl-9-thioxanthone, 4-Isopropyl-9H-thioxanthen-9-one, 4-Isopropylthioxanthone
IUPAC Name 4-propan-2-ylthioxanthen-9-one
SMILES CC(C)c1cccc2C(=O)c3ccccc3Sc12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H14OS/c1-10(2)11-7-5-8-13-15(17)12-6-3-4-9-14(12)18-16(11)13/h3-10H,1-2H3
Formula Weight 254.0765
Chemical Name or Material 4-Isopropylthioxantone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.