missing translation for 'onlineSavingsMsg'
Learn More

4-Keto 9-cis Retinoic Acid, TRC

Product Code. 30407299
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407299 2.5 mg 2.5mg
30394962 5 mg 5mg
30393949 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407299 Supplier Toronto Research Chemicals Supplier No. TRCK2049602.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150737-18-1
Molecular Formula C20 H26 O3
Molecular Weight (g/mol) 314.42
Synonym BAL10088, BAL0010088-000, BAL0004079-000, BAL4079, 9-cis retinoic acid, alitretinoin related
IUPAC Name (2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethyl-3-oxocyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
SMILES C\C(=C\C=C\C(=C\C(=O)O)\C)\C=C\C1=C(C)C(=O)CCC1(C)C

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C20H26O3/c1-14(7-6-8-15(2)13-19(22)23)9-10-17-16(3)18(21)11-12-20(17,4)5/h6-10,13H,11-12H2,1-5H3,(H,22,23)/b8-6+,10-9+,14-7-,15-13+
Formula Weight 314.1882
Chemical Name or Material 4-Keto 9-cis Retinoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.