missing translation for 'onlineSavingsMsg'
Learn More

4-Mercapto-4-methyl-2-pentanol, 98%

Product Code. 15454787
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15454787 250 mg 250mg
15444787 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454787 Supplier Thermo Scientific Alfa Aesar Supplier No. H32107.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 31539-84-1 | C6H14OS | 134.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31539-84-1
Molecular Formula C6H14OS
Molecular Weight (g/mol) 134.237
MDL Number MFCD10688294
InChI Key FDBQLLMYSACLPB-UHFFFAOYSA-N
PubChem CID 6429308
IUPAC Name 4-methyl-4-sulfanylpentan-2-ol
SMILES CC(CC(C)(C)S)O

Specifications

Density 0.957
Boiling Point 40°C (0.6 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.471
Quantity 250 mg
Synonym 4-mercapto-4-methyl-2-pentanol, +/--4-mercapto-4-methyl-2-pentanol, 4-mercapto-4-methylpentan-2-ol, 2-pentanol, 4-mercapto-4-methyl, 2-pentanol,4-mercapto-4-methyl, fema no. 4158, 4-mercapto-4-methyl-2-pentanol, +/-, +/--4-mercapto-4-methyl-2-pentanol fhfi, 4-mercapto-4-methyl pentan-2-ol
Formula Weight 134.24
Percent Purity 98%
Chemical Name or Material 4-Mercapto-4-methyl-2-pentanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.