missing translation for 'onlineSavingsMsg'
Learn More

4-Mercapto-4-methyl-2-pentanone, 98%

Product Code. 15474307
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474307 25 g 25g
15454307 1 g 1g
15464307 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15474307 Supplier Thermo Scientific Alfa Aesar Supplier No. H31693.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19872-52-7 | C6H12OS | 132.221 g/mol

4-Mercapto-4-methyl-2-pentanone is one of the most strongly contributing odorants in the volatile fraction of a Japanese green tea (sen-cha) infusion, and on the basis of the results of an aroma extract dilution analysis. 4-mercapto-4-methyl-2-pentanone in the grapefruit/sulfury group was found in reconstituted concentrate after thermal concentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Mercapto-4-methyl-2-pentanone is one of the most strongly contributing odorants in the volatile fraction of a Japanese green tea (sen-cha) infusion, and on the basis of the results of an aroma extract dilution analysis. 4-mercapto-4-methyl-2-pentanone in the grapefruit/sulfury group was found in reconstituted concentrate after thermal concentration.

Solubility
Soluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19872-52-7
Molecular Formula C6H12OS
Molecular Weight (g/mol) 132.221
MDL Number MFCD00085208
InChI Key QRNZMFDCKKEPSX-UHFFFAOYSA-N
Synonym 4-mercapto-4-methyl-2-pentanone, 4-mercapto-4-methylpentan-2-one, 2-pentanone, 4-mercapto-4-methyl, 4-methyl-4-thiolpentan-2-one, 4-methyl-4-mercapto-2-pentanone, 4-methyl-4-mercaptopentan-2-one, unii-9524rg5zql, 4-methyl-4-sulfanyl-2-pentanone, 4-sulfanyl-4-methylpentan-2-one, 4-sulphanyl-4-methylpentan-2-one
PubChem CID 88290
ChEBI CHEBI:77856
IUPAC Name 4-methyl-4-sulfanylpentan-2-one
SMILES CC(=O)CC(C)(C)S

Specifications

Density 0.961
Boiling Point 174°C
Flash Point 54°C (129°F)
Odor Unpleasant
Refractive Index 1.462
Quantity 25 g
UN Number UN3336
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 132.23
Percent Purity 98%
Chemical Name or Material 4-Mercapto-4-methyl-2-pentanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.