missing translation for 'onlineSavingsMsg'
Learn More

4-Mercapto-4-methyl-2-pentanone, TRC

Product Code. 30483233
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483233 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483233 Supplier Toronto Research Chemicals Supplier No. TRCM257598500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 4-Mercapto-4-methyl-2-pentanone
CAS 19872-52-7
Molecular Formula C6H12SO
Quantity 500 mg
Synonym 2-Pentanone, 4-mercapto-4-methyl- (6CI, 8CI, 9CI, ACI), 4-Mercapto-4-methyl-2-pentanone (ACI), 2-Mercapto-2-methylpentan-4-one, 4-Methyl-4-mercapto-2-pentanone, 4-Methyl-4-sulfanylpentan-2-one, 4-Sulfanyl-4-methylpentan-2-one, 4MMP
InChI Formula InChI=1S/C6H12OS/c1-5(7)4-6(2,3)8/h8H,4H2,1-3H3
SMILES CC(=O)CC(C)(C)S
IUPAC Name 4-methyl-4-sulfanylpentan-2-one
Molecular Weight (g/mol) 132.22
Formula Weight 132.0609
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.