missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-1,2-benzisoxazol-3-amine, 97%, Thermo Scientific™

Product Code. 10487622
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10487622 250 mg Amber Glass Bottle 250mg
10591122 1 g Amber Glass Bottle 1g
10526124 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10487622 Supplier Thermo Scientific Maybridge Supplier No. MO00345CB
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 177995-40-3
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.164
MDL Number MFCD03407354
InChI Key YFNIGSJHKGKXEQ-UHFFFAOYSA-N
Synonym 4-methoxy-1,2-benzisoxazol-3-amine, 4-methoxybenzo d isoxazol-3-amine, 4-methoxy-benzo d isoxazol-3-ylamine, 3-amino-4-methoxy-1,2-benzisoxazole, 1,2-benzisoxazol-3-amine, 4-methoxy, maybridge3_004235, 4-methoxybenzo d isoxazol-3-ylamine, 4-methoxy-1,2-benzisoazole-3-amine, 4-methoxybenzo d isoxazole-3-ylamine, 4-methoxy-1,2-benzisoxazole-3-amine
PubChem CID 2779751
IUPAC Name 4-methoxy-1,2-benzoxazol-3-amine
SMILES COC1=CC=CC2=C1C(=NO2)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material 4-Methoxy-1, 2-benzisoxazol-3-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.