missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-nitrobenzonitrile, TRC

Product Code. 30482865
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482865 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482865 Supplier Toronto Research Chemicals Supplier No. TRCM327008100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 4-Methoxy-3-nitrobenzonitrile
CAS 33224-23-6
Molecular Formula C8H6N2O3
Quantity 100 mg
Synonym 3-Nitro-4-Methoxybenzonitrile
InChI Formula InChI=1S/C8H6N2O3/c1-13-8-3-2-6(5-9)4-7(8)10(11)12/h2-4H,1H3
SMILES COC1=C([N+]([O-])=O)C=C(C#N)C=C1
IUPAC Name 4-methoxy-3-nitrobenzonitrile
Molecular Weight (g/mol) 178.15
Formula Weight 178.0378
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.