missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-d3-benzoic Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | 4-Methoxybenzoic Acid D3 (methoxy-D3) |
| CAS | 27914-54-1 |
| Molecular Formula | C8H5D3O3 |
| Quantity | 0.25 g |
| Synonym | p-Anisic-α,α,α-d3 Acid, 4-(Trideuteromethoxy)benzoic Acid |
| InChI Formula | InChI=1 S/C8H8O3/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5 H,1H3,(H,9,10)/i1D3 |
| SMILES | O=C(O)C1=CC=C(OC([2 H])([2 H])[2 H])C=C1 |
| IUPAC Name | 4-(methoxy-d3)benzoic acid |
| Molecular Weight (g/mol) | 155.17 |
| Formula Weight | 155.07 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction