missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy Retinoic Acid, TRC

Product Code. 30477987
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
Unit Size:
10mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477987 2.5 mg 2.5mg
30477041 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477987 Supplier Toronto Research Chemicals Supplier No. TRCM2678002.5MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81121-20-2
Molecular Formula C21 H30 O3
Molecular Weight (g/mol) 330.46
Synonym 4-Methoxyretinoic acid (9CI), (±)-4-Methoxyretinoic acid
IUPAC Name (2E,4E,6E,8E)-9-(3-methoxy-2,6,6-trimethylcyclohexen-1-yl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid
SMILES COC1CCC(C)(C)C(=C1C)\C=C\C(=C\C=C\C(=C\C(=O)O)\C)\C

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C21H30O3/c1-15(8-7-9-16(2)14-20(22)23)10-11-18-17(3)19(24-6)12-13-21(18,4)5/h7-11,14,19H,12-13H2,1-6H3,(H,22,23)/b9-7+,11-10+,15-8+,16-14+
Formula Weight 330.2195
Chemical Name or Material 4-Methoxy Retinoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.