missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxyacetophenone, 99%

Product Code. p-7081264
Change view
Click to view available options
Quantity:
25 g
250 g
1000 g
Unit Size:
1000g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314709 25 g 25g
11439093 250 g 250g
11429093 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11314709 Supplier Thermo Scientific Alfa Aesar Supplier No. A11162.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-06-1 | C9H10O2 | 150.18 g/mol

4'-Methoxyacetophenone is used as a component of perfumes and as chemical intermediate in the manufacture of pharmaceuticals, resins, flavouring agents. And also used to study ruthenium catalyzed step growth copolymerization of 4?-methoxyacetophenone with α,ω-dienes to yield copolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Methoxyacetophenone is used as a component of perfumes and as chemical intermediate in the manufacture of pharmaceuticals, resins, flavouring agents. And also used to study ruthenium catalyzed step growth copolymerization of 4′-methoxyacetophenone with α,ω-dienes to yield copolymers.

Solubility
Insoluble in water.

Notes
Light Sensitive, Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-06-1
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00008745
InChI Key NTPLXRHDUXRPNE-UHFFFAOYSA-N
Synonym 4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl
PubChem CID 7476
ChEBI CHEBI:86567
IUPAC Name 1-(4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 36°C to 40°C
Density 1.08
Boiling Point 258°C to 260°C
Flash Point 138°C (280°F)
Odor Characteristic
Quantity 25 g
Beilstein 742313
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material 4'-Methoxyacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.