missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzenemethanamine, TRC

Product Code. 30473169
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30473169 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30473169

Brand: Toronto Research Chemicals TRCM26091025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methoxybenzenemethanamine
CAS 2393-23-9
Molecular Formula C8H11NO
Quantity 25 g
Synonym (4-Methoxyphenyl)methanamine, 1-(4-Methoxyphenyl)methanamine, 1-Aminomethyl-4-methoxybenzene, 4-Methoxybenzenemethanamine, 4-Methoxybenzylamine, 4-Methoxyphenylmethylamine, 4-Methyloxybenzylamine, Anisylamine, NSC 9269, [[4-(Methyloxy)phenyl]methyl]amine, p-Methoxybenzylamine, para-Methoxybenzylamine
InChI Formula InChI=1S/C8H11NO/c1-10-8-4-2-7(6-9)3-5-8/h2-5H,6,9H2,1H3
SMILES COc1ccc(CN)cc1
Molecular Weight (g/mol) 137.18
Formula Weight 137.0841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.